Fisher BioReagents BP365500 Potassium Carbonate Anhydrous (White Granular Powder) (500g)

Representative image only.
Fisher BioReagents Mfr # BP365500 – Item # EW-88066-91

$90.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaK2CO3
  • ClassReagent Salts
  • ChemistrySolutions
  • FormPowder
  • SynonymsDipotassium Carbonate
  • Weight (g)500
  • Formula Weight138.21
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number584087
  • DescriptionPotassium Carbonate Anhydrous (White Granular Powder) (500g)

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat