Fisher BioReagents Potassium Iodide, 500 g

Representative image only.
Fisher BioReagents Mfr # BP367500 – Item # EW-88066-94
Quality and consistency your lab can count on
  • This potassium salt is used in buffers for molecular biology and cell culture applications

$553.50USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaKI
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)500
  • Formula Weight166.00
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number7681-11-0
  • Boiling Point C (° C)1330
  • Melting Point C (° C)680
  • DescriptionPotassium Iodide, 500 g
IrritantStorage Code Gray

Key Features

  • This potassium salt is used in buffers for molecular biology and cell culture applications

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat