Fisher BioReagents L-Cysteine Hydrochloride Monohydrate, White Crystals; 100 g

Representative image only.
Fisher BioReagents Mfr # BP376100 – Item # EW-88067-01
Reagents and solutions to meet your lab and research needs
  • This amino acid is suitable for use in tissue culture systems requiring additives
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC3H7NO2S•HCl•H2O
  • ClassSolids
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)100
  • Formula Weight175.64
  • PackagingGlass Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number7048-04-6
  • Melting Point C (° C)175
  • DescriptionL-Cysteine Hydrochloride Monohydrate, White Crystals; 100 g
IrritantStorage Code Gray

Key Features

  • This amino acid is suitable for use in tissue culture systems requiring additives

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat