Fisher BioReagents BP386100 L-Lysine Hydrochloride (White Powder) (100g)

Representative image only.
Fisher BioReagents Mfr # BP386100 – Item # EW-88067-12
  • Assay - 98.5-100.5%
  • Specific Rotation [α]&alpha20D; (c=8, 6N HCl) - +20.7° to +21.5° (+25.9° to 26.9° calc. as free base)
  • State of Solution - ≥98.0% transmittance
  • Chloride - 19.12-19.51%
  • Ammonium - ≤0.02%

$71.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC6H14N2O2•HCl
  • ClassSolids
  • ChemistrySolutions
  • FormPowder
  • Weight (g)100
  • Formula Weight182.65
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number657272
  • DescriptionL-Lysine Hydrochloride (White Powder) (100g)

Key Features

  • Assay - 98.5-100.5%
  • Specific Rotation [α]&alpha20D; (c=8, 6N HCl) - +20.7° to +21.5° (+25.9° to 26.9° calc. as free base)
  • State of Solution - ≥98.0% transmittance
  • Chloride - 19.12-19.51%
  • Ammonium - ≤0.02%
  • Heavy Metals (as Pb) - ≤10ppm
  • Arsenic - ≤1ppm
  • Other Amino Acids - Chromatographically not detectable
  • Loss on Drying at 105°C - ≤0.40%
  • Residue on Ignition (sulfated) - ≤0.10%

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat