Fisher BioReagents BP431100 Polyvinyl Pyrrolidone; 100 g

Representative image only.
Fisher Chemical Mfr # BP431100 – Item # EW-88067-32
Reagents and solutions to meet your lab and research needs
  • Assay (dry basis) - ≥95%
  • pH (5% in H2O at Room Temperature) - 3.0 to 7.0
  • Ash - ≤0.05%
  • Arsenic - ≤0.0002%
  • Heavy Metals (as Pb) - ≤0.001%
  • Viscosity at 23°C - 1.2 to 1.3 Centistokes
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Empirical Formula(C6H9NO)n
  • ClassSolids
  • ChemistrySolutions
  • FormSolid
  • SynonymsPovidone, Polyvidone, PVP
  • Weight (g)100
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number9003-39-8
  • DescriptionPolyvinylpyrrolidone (100g) CAS 9003-39-8
Storage Code Gray

Key Features

  • Assay (dry basis) - ≥95%
  • pH (5% in H2O at Room Temperature) - 3.0 to 7.0
  • Ash - ≤0.05%
  • Arsenic - ≤0.0002%
  • Heavy Metals (as Pb) - ≤0.001%
  • Viscosity at 23°C - 1.2 to 1.3 Centistokes

More About this Item

Quality and consistency your lab can count on.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat