Fisher BioReagents n-Butyl Alcohol, 500 mL

Representative image only.
Fisher BioReagents Mfr # BP505500 – Item # EW-88067-50
Reagents and solutions to meet your lab and research needs
  • n-Butanol is used as an analytical reagent and solvent

$351.75USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaCH3CH2CH2CH2OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • Synonyms1-Butanol, n-Butyl Alcohol
  • FormLiquid
  • Volume (mL)500
  • Formula Weight74.12
  • PackagingGlass Bottle
  • DOT InformationButanols; Class 3, Packing Group III
  • Storage SymbolRed
  • Hazard SymbolFlammable, Harmful
  • UN NumberUN1120
  • CAS Number71-36-3
  • Boiling Point C (° C)117.6
  • Melting Point C (° C)-89
  • Descriptionn-Butyl Alcohol, 500 mL
HarmfulStorage Code Red

Key Features

  • n-Butanol is used as an analytical reagent and solvent

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat