Fisher BioReagents BP541100 Manganese Chloride, 1.0M Soln. (Clear, Lt.-Pink Soln./Molecular Biology) (100mL)

Representative image only.
Fisher Chemical Mfr # BP541100 – Item # EW-88067-72
Quality and consistency your lab can count on
  • Concentration - 1.0M±0.01
  • Heavy Metals - ≤5.0ppm
  • DNase - Not detected
  • RNase - Not detected
  • Protease - Not detected

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeMolecular Biology
  • ClassReagent Salts
  • ChemistrySolutions
  • FormLiquid
  • SynonymsManganese Dichloride; Manganous Chloride
  • Volume (mL)100
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolCorrosive, Harmful
  • CAS Number7773-01-5
  • DescriptionManganese Chloride, 1.0M Soln. (Clear, Lt.-Pink Soln./Molecular Biology) (100mL)
HarmfulStorage Code Gray

Key Features

  • Concentration - 1.0M±0.01
  • Heavy Metals - ≤5.0ppm
  • DNase - Not detected
  • RNase - Not detected
  • Protease - Not detected

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat