Fisher BioReagents BP6755 Bovine Serum Albumin Solution (20mg/ mL) (5 mL)

Representative image only.
Fisher BioReagents Mfr # BP6755 – Item # EW-88068-34
Ideal for use in stabilizing enzymes during DNA digestion
  • Total Protein (Bradford) - 18-22mg/mL
  • Optical Absorbance (dilution, 1:40 in water) Electrophoresis - to pass test
  • Free of DNase, RNase, and protease

SPECS & DESCRIPTION

Specifications & Description

  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)5
  • PackagingAmber Glass
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number9048-46-8 and 26628-22-8
  • DescriptionBovine Serum Albumin Solution (20mg/ mL) (5 mL)
IrritantStorage Code Gray

Key Features

  • Total Protein (Bradford) - 18-22mg/mL
  • Optical Absorbance (dilution, 1:40 in water) Electrophoresis - to pass test
  • Free of DNase, RNase, and protease

More About this Item

The Bovine Serum Albumin (BSA) solution can be used as an additive or nutrient in cell and microbial culture. You can also use it to determine quantity of unknow proteins in protein assays.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat