Fisher BioReagents BP91050 Streptomycin Sulfate, Powder; 50 g

Representative image only.
Acros Organics Mfr # BP91050 – Item # EW-88068-51
Reagents and solutions to meet your lab and research needs
  • pH of a 20% Solution - 4.5 to 7.0
  • Loss on Drying at 105°C - ≤5%

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC42H84N14O36S3
  • Linear Formula(C21H39N7O12)2•3H2SO4
  • ClassSolids
  • ChemistrySolutions
  • FormPowder
  • Weight (g)50
  • Formula Weight1457.58
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolToxic
  • CAS Number3810-74-0
  • DescriptionStreptomycin Sulfate (50g) streptomycin sulfate 50g
Storage Code Gray

Key Features

  • pH of a 20% Solution - 4.5 to 7.0
  • Loss on Drying at 105°C - ≤5%

More About this Item

Quality and consistency your lab can count on.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat