Fisher BioReagents Citric Acid Anhydrous, 500 g

Representative image only.
Fisher BioReagents Mfr # BP339500 – Item # EW-88065-79
Quality and consistency your lab can count on
  • Anhydrous citric acid is suitable for use in tissue culture systems requiring additives

$118.00USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC6H8O7
  • ClassReagent Acids and Bases
  • ChemistryAcids
  • FormSolid
  • Weight (g)500
  • Formula Weight192.13
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number77-92-9
  • Melting Point C (° C)153
  • DescriptionCitric Acid Anhydrous, 500 g
IrritantStorage Code Gray

Key Features

  • Anhydrous citric acid is suitable for use in tissue culture systems requiring additives

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat