Fisher Chemical A18500 Acetone (Certified ACS) (500 mL) CAS 67-64-1

Representative image only.
Fisher Chemical Mfr # A18500 – Item # EW-88000-68
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.5%
  • Color (APHA)—≤10
  • Isopropyl Alcohol—≤0.05%
  • Density at 25°C—≤0.7857g/mL
  • Boiling Range (1 drop to dryness)—≤0.8°C
  • Boiling Point—56.1° ±0.1°C
  • Residue after Evaporation—≤0.001%
  • Solubility in H2O—To pass test
  • Aldehyde (as HCHO)—≤0.002%
  • Titratable Acid—≤0.0003mEq/g
  • Titratable Base—≤0.0006mEq/g
  • Methanol—≤0.05%
  • Substances Reducing Permanganate—To pass test
  • IR Transmittance—To pass test
  • Water—≤0.5%
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaC3H6O
  • Linear FormulaCH3COCH3
  • ClassReagent Solvents
  • ChemistryAnalytical Reagents
  • FormLiquid
  • Synonyms2-Propanone, Pyroacetic Ether, 2-Propanone, Dimethyl Ketone
  • Volume (mL)500
  • Formula Weight58.08
  • PackagingAmber Glass
  • DOT InformationFlammable Liquid; Class 3
  • Storage SymbolRed
  • Hazard SymbolFlammable, Irritant
  • CAS Number67-64-1
  • UN NumberUN1090
  • DescriptionAcetone (Certified ACS) (500 mL)
FlammableIrritantStorage Code Red

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.5%
  • Color (APHA)—≤10
  • Isopropyl Alcohol—≤0.05%
  • Density at 25°C—≤0.7857g/mL
  • Boiling Range (1 drop to dryness)—≤0.8°C
  • Boiling Point—56.1° ±0.1°C
  • Residue after Evaporation—≤0.001%
  • Solubility in H2O—To pass test
  • Aldehyde (as HCHO)—≤0.002%
  • Titratable Acid—≤0.0003mEq/g
  • Titratable Base—≤0.0006mEq/g
  • Methanol—≤0.05%
  • Substances Reducing Permanganate—To pass test
  • IR Transmittance—To pass test
  • Water—≤0.5%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat