Fisher Chemical A200C212 Nitric Acid R Acs 6x21/2l/cs

Representative image only.
Fisher Chemical Mfr # A200C212 – Item # EW-88050-69
Quality and consistency your lab can count on

PRODUCT OPTIONS

$577.00USD / Case

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaHNO3
  • Linear FormulaHNO3
  • ClassReagent Acids and Bases
  • ChemistryAcids;Inorganics
  • FormLiquid
  • Volume (mL)2500
  • Formula Weight63.01
  • PackagingGlass bottle
  • DOT InformationNitric Acid; Class 8 (5.1), Packing Group II
  • Hazard SymbolCorrosive, Oxidizing, Toxic
  • CAS Number7697-37-2 and 7732-18-5
  • UN NumberUN2031
  • DescriptionNitric Acid R ACS Plus (6x2.5 L)

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat