Fisher Chemical Phosphorus Pentoxide, Certified ACS Grade, 500 g

Representative image only.
Fisher Chemical Mfr # A245500 – Item # EW-88019-62
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.0%
  • Insoluble Matter — ≤0.01%
  • Phosphorus Trioxide — To pass test (about 0.02%)
  • Ammonium — ≤0.005%
  • Heavy Metals (as Pb) — ≤0.005%

PRODUCT OPTIONS

$305.75USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaP2O5
  • ClassSolids
  • ChemistryAnalytical Reagents
  • FormSolid
  • SynonymsPhosphoric Anhydride, Phosphorus oxide
  • Weight (g)500
  • Formula Weight141.94
  • PackagingGlass Bottle
  • DOT InformationPhosphorus Pentoxide; Class 8, Packing Group II
  • Storage SymbolWhite
  • Hazard SymbolCorrosive
  • CAS Number1314-56-3
  • UN NumberUN1807
  • Boiling Point C (° C)360
  • Melting Point C (° C)340
  • DescriptionPhosphorus Pentoxide, Certified ACS Grade, 500 g
CorrosiveStorage Code White

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.0%
  • Insoluble Matter — ≤0.01%
  • Phosphorus Trioxide — To pass test (about 0.02%)
  • Ammonium — ≤0.005%
  • Heavy Metals (as Pb) — ≤0.005%

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat