Fisher Chemical Propionic Acid, Certified Grade, 500 mL

Representative image only.
Fisher Chemical Mfr # A258500 – Item # EW-88021-68
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Color (APHA) — ≤15
  • Boiling Range — 139°-142°C
  • Density at 25°C — 0.986-0.990g/mL
  • Residue after Evaporation — ≤0.01%
  • Water — ≤0.25%

$119.75USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeCertified
  • Empirical FormulaCH3CH2COOH
  • ClassReagent Acids and Bases
  • ChemistryAcids
  • FormLiquid
  • SynonymsMethyl Acetic Acid, Propanoic Acid, Ethyl Carbonic Acid
  • Volume (mL)500
  • Formula Weight74.08
  • PackagingGlass Bottle
  • DOT InformationPropionic Acid; Class 8, Packing Group III
  • Storage SymbolRed
  • Hazard SymbolCorrosive, Flammable, Toxic
  • UN NumberUN1848
  • CAS Number79-09-4
  • Boiling Point C (° C)3
  • Melting Point C (° C)-22
  • DescriptionPropionic Acid, Certified Grade, 500 mL
CorrosiveStorage Code Red

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Color (APHA) — ≤15
  • Boiling Range — 139°-142°C
  • Density at 25°C — 0.986-0.990g/mL
  • Residue after Evaporation — ≤0.01%
  • Water — ≤0.25%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat