Fisher Chemical Sulfuric Acid Solution, Certified ACS PlusGrade, 500 mL

Representative image only.
Fisher Chemical Mfr # A300500 – Item # EW-88030-12
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay — 95.0-98.0%
  • Color (APHA) — ¡Ü10
  • Residue after Ignition — ¡Ü3ppm
  • Chloride — ¡Ü0.1ppm
  • Nitrate — ¡Ü0.2ppm
  • Ammonium — ¡Ü1ppm
  • Substances Reducing Permanganate (as SO2) — To pass test (about 2ppm)
  • Heavy Metals — ¡Ü0.8ppm
  • Phosphate — ¡Ü0.5ppm
  • Aluminum — ¡Ü0.2ppm
  • Arsenic — ¡Ü0.004ppm
  • Boron — ¡Ü0.01ppm
  • Calcium — ¡Ü0.3ppm
  • Chromium — ¡Ü0.2ppm
  • Copper — ¡Ü0.1ppm
  • Gold — ¡Ü0.3ppm
  • Iron — ¡Ü0.2ppm
  • Lead — ¡Ü0.3ppm
  • Magnesium — ¡Ü0.3ppm
  • Manganese — ¡Ü0.2ppm
  • Mercury — ¡Ü5ppb
  • Nickel — ¡Ü0.1ppm
  • Potassium — ¡Ü0.3ppm
  • Sodium — ¡Ü0.3ppm
  • Tin — ¡Ü0.2ppm
  • Titanium — ¡Ü0.3ppm
  • Zinc — ¡Ü0.2ppm

PRODUCT OPTIONS

$204.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration95 to 98%
  • Purity GradeCertified ACS Plus
  • Empirical FormulaH2SO4
  • MatrixWater
  • ClassReagent Acids and Bases
  • ChemistryAcids
  • FormLiquid
  • SynonymsHygrogen sulfate, Oil of vitriol, Vitriol brown oil, Mattling acid, Battery acid, Sulphuric acid, Electrolyte acid, Dihydrogen sulfate, Spirt of sulphur, Chamber acid
  • Volume (mL)500
  • Formula Weight98.08
  • PackagingGlass Bottle
  • DOT InformationSulfuric acid; Class 8, Packing Group II
  • Storage SymbolWhite
  • Hazard SymbolCorrosive
  • CAS Number7664-93-9
  • UN NumberUN1830
  • Boiling Point C (° C)290
  • Melting Point C (° C)10
  • DescriptionSulfuric Acid Solution, Certified ACS PlusGrade, 500 mL
CorrosiveStorage Code White

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — 95.0-98.0%
  • Color (APHA) — ¡Ü10
  • Residue after Ignition — ¡Ü3ppm
  • Chloride — ¡Ü0.1ppm
  • Nitrate — ¡Ü0.2ppm
  • Ammonium — ¡Ü1ppm
  • Substances Reducing Permanganate (as SO2) — To pass test (about 2ppm)
  • Heavy Metals — ¡Ü0.8ppm
  • Phosphate — ¡Ü0.5ppm
  • Aluminum — ¡Ü0.2ppm
  • Arsenic — ¡Ü0.004ppm
  • Boron — ¡Ü0.01ppm
  • Calcium — ¡Ü0.3ppm
  • Chromium — ¡Ü0.2ppm
  • Copper — ¡Ü0.1ppm
  • Gold — ¡Ü0.3ppm
  • Iron — ¡Ü0.2ppm
  • Lead — ¡Ü0.3ppm
  • Magnesium — ¡Ü0.3ppm
  • Manganese — ¡Ü0.2ppm
  • Mercury — ¡Ü5ppb
  • Nickel — ¡Ü0.1ppm
  • Potassium — ¡Ü0.3ppm
  • Sodium — ¡Ü0.3ppm
  • Tin — ¡Ü0.2ppm
  • Titanium — ¡Ü0.3ppm
  • Zinc — ¡Ü0.2ppm

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat