Fisher Chemical Acetic Acid, Glacial, Certified ACS Grade, 2.5 L; 6/Cs

Representative image only.
Fisher Chemical Mfr # A38C212 – Item # EW-88051-47
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

PRODUCT OPTIONS

$752.50USD / Case of 6

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeCertified ACS
  • Empirical FormulaCH3COOH
  • ClassReagent Acids and Bases
  • ChemistryAcids
  • SynonymsMethanecarboxylic Acid, Vinegar Acid
  • FormLiquid
  • Volume (mL)2500
  • Formula Weight60.05
  • PackagingGlass Bottle
  • DOT InformationAcetic acid, glacial; Class 8 (3), Packing Group II
  • Storage SymbolRed
  • Hazard SymbolCorrosive
  • CAS Number64-19-7
  • UN NumberUN2789
  • Boiling Point C (° C)117
  • Melting Point C (° C)16
  • DescriptionAcetic Acid, Glacial, Certified ACS Grade, 2.5 L; 6/Cs

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat