Fisher Chemical A399500 1-Butanol, Certified ACS; 500 mL

Representative image only.
Fisher Chemical Mfr # A399500 – Item # EW-88003-86
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.4%
  • Carbonyl Compounds (as Butyraldehyde) — ≤0.01%
  • Color (APHA) — ≤10
  • Titratable Acid — ≤0.0008mEq/g
  • Residue after Evaporation — ≤0.005%
  • Water — ≤0.1%
  • Fluorescent Impurities (as Quinine Sulfate) — To pass test (≤0.02μg/mL)
  • Butyl Ether (by GC) — ≤0.2%
  • Boiling Range (1 drop to dryness) — ≤1.5°C
  • Boiling Point — 117.7°C ± 0.1°C

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaC4H10O
  • Linear FormulaCH3CH2CH2CH2OH
  • ClassReagent Solvents
  • ChemistryAnalytical Reagents
  • Synonymsn-Butyl Alcohol
  • FormLiquid
  • Volume (mL)500
  • Formula Weight74.12
  • PackagingAmber Glass
  • DOT InformationFlammable Liquid; Class 3, Packing Group III
  • Storage SymbolRed
  • Hazard SymbolHarmful
  • UN NumberUN1120
  • CAS Number71363
  • Description1-Butanol (Certified ACS) (500 mL) CAS 71-36-3
HarmfulStorage Code Red

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.4%
  • Carbonyl Compounds (as Butyraldehyde) — ≤0.01%
  • Color (APHA) — ≤10
  • Titratable Acid — ≤0.0008mEq/g
  • Residue after Evaporation — ≤0.005%
  • Water — ≤0.1%
  • Fluorescent Impurities (as Quinine Sulfate) — To pass test (≤0.02μg/mL)
  • Butyl Ether (by GC) — ≤0.2%
  • Boiling Range (1 drop to dryness) — ≤1.5°C
  • Boiling Point — 117.7°C ± 0.1°C

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat