Fisher Chemical Isopropanol, Certified ACS Reagent Grade, 20 L

Representative image only.
Fisher Chemical Mfr # A41620 – Item # EW-88052-14
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

PRODUCT OPTIONS

$710.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1
  • Purity GradeCertified ACS Reagent
  • Empirical Formula(CH3)2CHOH
  • MatrixNone
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • Synonyms2-Propanol, IPA, Isopropyl alcohol
  • Volume (mL)20000
  • Formula Weight60.10
  • PackagingSteel Pail
  • DOT InformationIsopropanol; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable, Irritant
  • CAS Number67-63-0
  • UN NumberUN1219
  • Boiling Point C (° C)81
  • Melting Point C (° C)-89.5
  • DescriptionIsopropanol, Certified ACS Reagent Grade, 20 L

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat