Fisher Chemical A43420 Methanol Acs In Spec 20l Drum

Representative image only.
Fisher Chemical Mfr # A43420 – Item # EW-88052-59
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaCH4O
  • Linear FormulaCH3OH
  • ClassReagent Solvents
  • ChemistrySolvents;Analytical Reagents
  • FormLiquid
  • SynonymsWood Alcohol, Methyl Alcohol
  • Volume (mL)20000
  • Formula Weight32.04
  • PackagingLined Steel Drum
  • DOT InformationMethanol; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable, Irritant
  • CAS Number67-56-1
  • UN NumberUN1230
  • DescriptionMethanol ACS In Spec (20 L) Drum

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat