Fisher Chemical Methanol, HPLC Grade, Safe-Cote™ Amber Glass Bottle, 4 L

Representative image only.
Fisher Chemical Mfr # A452SK4 – Item # EW-88015-16
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.9%
  • Color (APHA) — ≤5
  • Fluorescence Background (as Quinine Sulfate) — To pass test
  • Refractive Index at 25°C — 1.3260-1.3300
  • Residue after Evaporation — ≤3ppm
  • Solubility in H2O — To pass test
  • Substances Darkened by H2SO4 — To pass test
  • Substances Reducing Permanganate — To pass test
  • Water — ≤0.1%
  • Titratable Acid — ≤0.0003mEq/g
  • Titratable Base — ≤0.0002mEq/g
  • LC Gradient Suitability — To pass test
  • Carbonyl Compounds (each of Acetone, Formaldehyde and Acetaldehyde) — ≤0.001%
  • Optical Absorbance at 205nm — ≤1.00
  • at 220nm — ≤0.30
  • at 230nm — ≤0.15
  • at 254nm — ≤0.025
  • Submicron Filtered.

PRODUCT OPTIONS

$227.75USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeHPLC
  • Empirical FormulaCH3OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • SynonymsWood Alcohol, Methyl Alcohol
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight32.04
  • PackagingSafe-Cote™ Amber Glass Bottle
  • DOT InformationMethanol; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable, Toxic
  • UN NumberUN1230
  • CAS Number67-56-1
  • Boiling Point C (° C)64.7
  • Melting Point C (° C)-98
  • DescriptionMethanol, HPLC Grade, Safe-Cote™ Amber Glass Bottle, 4 L
FlammableStorage Code RedToxic

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.9%
  • Color (APHA) — ≤5
  • Fluorescence Background (as Quinine Sulfate) — To pass test
  • Refractive Index at 25°C — 1.3260-1.3300
  • Residue after Evaporation — ≤3ppm
  • Solubility in H2O — To pass test
  • Substances Darkened by H2SO4 — To pass test
  • Substances Reducing Permanganate — To pass test
  • Water — ≤0.1%
  • Titratable Acid — ≤0.0003mEq/g
  • Titratable Base — ≤0.0002mEq/g
  • LC Gradient Suitability — To pass test
  • Carbonyl Compounds (each of Acetone, Formaldehyde and Acetaldehyde) — ≤0.001%
  • Optical Absorbance at 205nm — ≤1.00
  • at 220nm — ≤0.30
  • at 230nm — ≤0.15
  • at 254nm — ≤0.025
  • Submicron Filtered.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat