Fisher Chemical Methanol, Residue Analysis Grade, Purge and Trap Suitable for Volatile Organic, Amber Glass Bottle, 500 mL

Representative image only.
Fisher Chemical Mfr # A453500 – Item # EW-88015-02
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay (by GC) — ≥99.9%
  • Residue after Evaporation — ≤1ppm
  • Water — ≤0.1%
  • Other Volatile Impurities — ≤10ppb
  • Titratable Acid — ≤0.0003mEq/g
  • Titratable Base — ≤0.0002mEq/g
  • Packaged under Nitrogen, Submicron Filtered.

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeResidue Analysis
  • Empirical FormulaCH3OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • SynonymsWood Alcohol, Methyl Alcohol
  • FormLiquid
  • Volume (mL)500
  • Formula Weight32.04
  • PackagingAmber Glass Bottle
  • DOT InformationMethanol; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable, Toxic
  • UN NumberUN1230
  • CAS Number67-56-1
  • Boiling Point C (° C)64.7
  • Melting Point C (° C)-98
  • DescriptionMethanol, Residue Analysis Grade, Purge and Trap Suitable for Volatile Organic, Amber Glass Bottle, 500 mL
FlammableStorage Code RedToxic

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (by GC) — ≥99.9%
  • Residue after Evaporation — ≤1ppm
  • Water — ≤0.1%
  • Other Volatile Impurities — ≤10ppb
  • Titratable Acid — ≤0.0003mEq/g
  • Titratable Base — ≤0.0002mEq/g
  • Packaged under Nitrogen, Submicron Filtered.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat