Fisher Chemical A467500 Nitric Acid (OPTIMA*) (500 mL) CAS 7697-37-2

Representative image only.
Fisher Chemical Mfr # A467500 – Item # EW-88017-62
Quality and consistency your lab can count on

PRODUCT OPTIONS

$840.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeOPTIMA
  • Empirical FormulaHNO3
  • Linear FormulaHNO3
  • ClassReagent Acids and Bases
  • ChemistryAcids;Inorganics
  • FormLiquid
  • Volume (mL)500
  • Formula Weight63.01
  • PackagingPTFE FEP
  • DOT InformationCorrosive; Class 8 (5.1), Packing Group II
  • Storage SymbolYellow
  • Hazard SymbolCorrosive, Oxidizing
  • UN NumberUN2031
  • CAS Number7697-37-2 and 7732-18-5
  • DescriptionNitric Acid (OPTIMA*) (500 mL) CAS 7697-37-2

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat