Fisher Chemical A484212 Sulfuric Acid (NF/FCC) (2.5 liter) CAS 7664-93-9

Representative image only.
Fisher Chemical Mfr # A484212 – Item # EW-88030-18
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay — 95.0-98.0%
  • Residue on Ignition — ≤0.005%
  • Reducing Substances — To pass test
  • Arsenic — ≤1ppm
  • Chloride — ≤0.005%
  • Iron — ≤0.02%
  • Lead — ≤5ppm
  • Nitrate — ≤10ppm
  • Selenium — ≤0.002%
  • Heavy Metals — ≤5ppm

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeFCC or NF
  • Empirical FormulaH2SO4
  • Linear FormulaH2SO4
  • ClassReagent Acids and Bases
  • ChemistryAcids;Inorganics
  • SynonymsHygrogen sulfate; Oil of vitriol; Vitriol brown oil; Mattling acid; Battery acid; Sulphuric acid; Electrolyte acid; Dihydrogen sulfate; Spirt of sulphur; Chamber acid
  • FormLiquid
  • Volume (mL)2500
  • Formula Weight98.08
  • PackagingGlass
  • DOT InformationCorrosive; Class 8, Packing Group II
  • Storage SymbolWhite
  • Hazard SymbolCorrosive
  • CAS Number7664-93-9
  • UN NumberUN1830
  • DescriptionSulfuric Acid (NF/FCC) (2.5 L)
CorrosiveStorage Code White

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — 95.0-98.0%
  • Residue on Ignition — ≤0.005%
  • Reducing Substances — To pass test
  • Arsenic — ≤1ppm
  • Chloride — ≤0.005%
  • Iron — ≤0.02%
  • Lead — ≤5ppm
  • Nitrate — ≤10ppm
  • Selenium — ≤0.002%
  • Heavy Metals — ≤5ppm

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat