Fisher Chemical BP11051 Methanol (Peroxide-free/Sequencing) (1 liter)

Representative image only.
Fisher Chemical Mfr # BP11051 – Item # EW-88015-42
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay — >=99.9%
  • IR — Conforms to standard
  • Color (APHA) — ≤5
  • Optical Absorbance at 205nm — ≤1.00
  • &tab; at 220nm — ≤0.30
  • &tab; at 230nm — ≤0.15
  • &tab; at 254nm — ≤0.025
  • Fluorescence Background — To pass test
  • Refractive Index at 25°C — 1.3260-1.3300
  • Titratable Acid — ≤0.0003mEq/g
  • Titratable Base — ≤0.0002mEq/g
  • Residue after Evaporation — ≤3ppm
  • Solubility in H2O — To pass test
  • Impurities Reactive with H2SO4 — Not detected
  • Water — ≤0.1%
  • Acetone — To pass test (about 0.001%)
  • Fluorescent Derivatizable Amines — To pass test
  • Peroxides — ≤0.001%
  • LC Gradient Suitability — To pass test
  • Substances Reducing Permanganate — To pass test

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeSequencing
  • Empirical FormulaCH4O
  • Linear FormulaCH3OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsWood Alcohol, Methyl Alcohol
  • Volume (mL)1000
  • Formula Weight32.04
  • PackagingAmber Glass
  • DOT InformationMethanol; Class 3 (6.1), Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable, Solvent, Toxic
  • UN NumberUN1230
  • CAS Number67-56-1
  • DescriptionMethanol (Peroxide-free/Sequencing) (1 L)
FlammableStorage Code RedToxic

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — >=99.9%
  • IR — Conforms to standard
  • Color (APHA) — ≤5
  • Optical Absorbance at 205nm — ≤1.00
  • &tab; at 220nm — ≤0.30
  • &tab; at 230nm — ≤0.15
  • &tab; at 254nm — ≤0.025
  • Fluorescence Background — To pass test
  • Refractive Index at 25°C — 1.3260-1.3300
  • Titratable Acid — ≤0.0003mEq/g
  • Titratable Base — ≤0.0002mEq/g
  • Residue after Evaporation — ≤3ppm
  • Solubility in H2O — To pass test
  • Impurities Reactive with H2SO4 — Not detected
  • Water — ≤0.1%
  • Acetone — To pass test (about 0.001%)
  • Fluorescent Derivatizable Amines — To pass test
  • Peroxides — ≤0.001%
  • LC Gradient Suitability — To pass test
  • Substances Reducing Permanganate — To pass test

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat