Fisher Chemical BP139892 SDS-PAGE Buffer Mix (Dry Powd. Mix of Tris/Glycine/SDS/Electrophor.) (92g)

Representative image only.
Fisher Chemical Mfr # BP139892 – Item # EW-88057-22
  • Solubility (buffer in final volume of 1 liter in H2O) - Clear solution
  • pH of a 1X Buffer at 25°C - 8.3 ±0.2
  • Electrophoresis - To pass test

SPECS & DESCRIPTION

Specifications & Description

  • ChemistrySolutions
  • Weight (g)92
  • CAS Number56-40-6 (Glycine), 77-86-1 (Tris), 151-21-3 (SDS)
  • DescriptionSDS-PAGE Buffer Mix (Dry Powd. Mix of Tris/Glycine/SDS/Electrophor.) (92g)
Storage Code Gray

Key Features

  • Solubility (buffer in final volume of 1 liter in H2O) - Clear solution
  • pH of a 1X Buffer at 25°C - 8.3 ±0.2
  • Electrophoresis - To pass test

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat