Fisher Chemical Dextran Sulfate Sodium Salt, 100 g

Representative image only.
Fisher Chemical Mfr # BP1585100 – Item # EW-88007-12
Quality and consistency your lab can count on
  • Dextran Sulfate accelerates the rate of hybridization of DNA or RNA bound to membranes and may be used in Southern and Northern blotting procedures
  • Sulfur Content — 17-20%
  • Moisture — ≤7.5%

SPECS & DESCRIPTION

Specifications & Description

  • FormSolid
  • Weight (g)100
  • PackagingGlass Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGrey
  • CAS Number9011-18-1
  • DescriptionDextran Sulfate Sodium Salt, 100 g
IrritantStorage Code Gray

Key Features

  • Dextran Sulfate accelerates the rate of hybridization of DNA or RNA bound to membranes and may be used in Southern and Northern blotting procedures
  • Sulfur Content — 17-20%
  • Moisture — ≤7.5%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat