Fisher Chemical BP1605100 Bovine Serum Albumin (Fraction V) (Cold-ethanol Precipitated) (100g)

Representative image only.
Fisher Chemical Mfr # BP1605100 – Item # EW-88003-25
  • Assay — ≥98%
  • Heavy Metals (as Pb) — ≤0.002%
  • Moisture — ≤5%
  • pH (1% Solution in 0.9% NaCl) — 7.0 ±0.3
  • Solubility (0.4g/10mL H2O) — Clear and haze-free

PRODUCT OPTIONS

$487.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ClassSolids
  • ChemistrySolutions
  • SynonymsBSA (Fraction V)
  • FormSolid
  • Weight (g)100
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number9048-46-8
  • DescriptionBovine Serum Albumin (Fraction V) (Cold-ethanol Precipitated) (100g)

Key Features

  • Assay — ≥98%
  • Heavy Metals (as Pb) — ≤0.002%
  • Moisture — ≤5%
  • pH (1% Solution in 0.9% NaCl) — 7.0 ±0.3
  • Solubility (0.4g/10mL H2O) — Clear and haze-free
  • Sulfated Ash — ≤2%

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat