Fisher Chemical Phenol, Saturated, pH 6.6/7.9, 400 mL

Representative image only.
Fisher Chemical Mfr # BP1750I400 – Item # EW-88022-16
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay — >99.0%
  • Optical Absorbance at 330nm — <0.2
  • &tab; at 405nm — <0.1
  • &tab; at 510nm — <0.1 pH — 6.6±0.2
  • pH (with buffer) — 7.9±0.2
  • DNase — Not detected
  • RNase — Not detected
  • Stability (4.0°C) — >1 year
  • Stability (with buffer) — 6 months

$417.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC6H6O
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)400
  • Formula Weight94.11
  • PackagingSafety Coated Amber Glass Poison Pack
  • DOT InformationPhenol Solutions; Class 6.1, Packing Group II
  • Storage SymbolWhite
  • Hazard SymbolFlammable, Toxic
  • UN NumberUN2821
  • CAS Number108-95-2
  • DescriptionPhenol, Saturated, pH 6.6/7.9, 400 mL
Storage Code WhiteToxic

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — >99.0%
  • Optical Absorbance at 330nm — <0.2
  • &tab; at 405nm — <0.1
  • &tab; at 510nm — <0.1 pH — 6.6±0.2
  • pH (with buffer) — 7.9±0.2
  • DNase — Not detected
  • RNase — Not detected
  • Stability (4.0°C) — >1 year
  • Stability (with buffer) — 6 months

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat