This item is now discontinued. Please browse similar items

Fisher Chemical BP2301 Ethylene Glycol (1 liter) CAS 107-21-1 (1,2-Ethanediol)

Representative image only.
Fisher Chemical Mfr # BP2301 – Item # EW-88058-48

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC2H6O2
  • Linear FormulaHOCH2CH2OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • Synonyms1,2-Ethanediol
  • Volume (mL)1000
  • Formula Weight62.07
  • CAS Number107-21-1
  • DescriptionEthylene Glycol (1 L)

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat