Fisher Chemical BP2482100 Ethylenediaminetetraacetic Acid (0.5 M Solution/pH 8.0) (100mL)

Representative image only.
Fisher Chemical Mfr # BP2482100 – Item # EW-88059-55
Quality and consistency your lab can count on
  • Chelating agent for metal ions
  • Minimizes metal ion impurities in reaction buffers

SPECS & DESCRIPTION

Specifications & Description

  • ChemistryAcids;Inorganics
  • Volume (mL)100
  • Formula Weight292.25
  • CAS Number60-00-4
  • DescriptionEthylenediaminetetraacetic Acid (0.5 M Solution/pH 8.0) (100mL)
IrritantStorage Code Gray

Key Features

  • Chelating agent for metal ions
  • Minimizes metal ion impurities in reaction buffers

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat