Fisher Chemical BP253010 Methyl alpha-D-mannopyranoside (10g)

Representative image only.
Fisher Chemical Mfr # BP253010 – Item # EW-88060-47
  • Assay - ≥99%
  • FTIR - Conforms to standard
  • Melting Point - 187-197°C

SPECS & DESCRIPTION

Specifications & Description

  • ChemistrySolutions
  • Weight (g)10
  • Formula Weight194.2
  • CAS Number617-04-9
  • DescriptionMethyl alpha-D-mannopyranoside (10g)
Storage Code Gray

Key Features

  • Assay - ≥99%
  • FTIR - Conforms to standard
  • Melting Point - 187-197°C

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat