Fisher Chemical BP351500 L-Ascorbic Acid (White Cryst. Powder) (500g)

Representative image only.
Fisher Chemical Mfr # BP351500 – Item # EW-88066-72

$160.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC6H8O6
  • ClassSolids
  • ChemistryAcids
  • FormPowder
  • SynonymsVitamin C
  • Weight (g)500
  • Formula Weight176.12
  • PackagingAmber Glass
  • Storage SymbolGray
  • CAS Number50817
  • DescriptionL-Ascorbic Acid (White Cryst. Powder) (500g)

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat