Fisher Chemical BP90825 Nalidixic Acid (25g) 8806850

Representative image only.
Fisher Chemical Mfr # BP90825 – Item # EW-88068-50
  • Assay - ≥98%
  • Heavy Metals (as Pb) - ≤0.002%
  • Melting Range - 225°-231°C
  • Loss on Drying at 105°C - ≤0.5%

SPECS & DESCRIPTION

Specifications & Description

  • ChemistryAcids
  • Weight (g)25
  • Formula Weight232.24
  • CAS Number389-08-2
  • DescriptionNalidixic Acid (25g) 8806850
HarmfulStorage Code Gray

Key Features

  • Assay - ≥98%
  • Heavy Metals (as Pb) - ≤0.002%
  • Melting Range - 225°-231°C
  • Loss on Drying at 105°C - ≤0.5%

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat