Fisher Chemical BP914100 Penicillin-G Potassium Salt (100 million units)

Representative image only.
Fisher Chemical Mfr # BP914100 – Item # EW-88068-53
  • Assay - ≥85%
  • pH of a 0.6% Solution - 5.5 to 7.5
  • Loss on Drying at 105°C - ≤1.0%

$130.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC16H17N2KO4S
  • ClassSolids
  • ChemistrySolutions
  • FormPowder
  • Weight (g)0.0001
  • Formula Weight372.47
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolFlammable, Harmful
  • CAS Number113-98-4
  • DescriptionPenicillin-G Potassium Salt (100 million units)

Key Features

  • Assay - ≥85%
  • pH of a 0.6% Solution - 5.5 to 7.5
  • Loss on Drying at 105°C - ≤1.0%

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat