Fisher Chemical C430500 Copper (Shot/Certified ACS) (500g) CAS 7440-50-8

Representative image only.
Fisher Chemical Mfr # C430500 – Item # EW-88006-00
Consistent, high quality organics and fine chemicals at great savings
  • Reagents and solutions to meet your lab and research needs
  • Assay — ¡Ý99.90%
  • Insoluble in Dilute HNO3 — ¡Ü0.020%
  • Antimony and Tin (as Sn) — ¡Ü0.01%
  • Arsenic — ¡Ü5ppm
  • Iron — ¡Ü0.005%
  • Lead — ¡Ü0.005%
  • Manganese — ¡Ü0.001%
  • Phosphorus — ¡Ü0.001%
  • Silver — ¡Ü20ppm
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaCu
  • ClassStandards
  • ChemistryAnalytical Reagents;Solutions
  • FormShot
  • SynonymsCopper shot and light turnings
  • Weight (g)500
  • Formula Weight63.55
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number7440-50-8
  • DescriptionCopper (Shot/Certified ACS) (500g) CAS 7440-50-8
Storage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ¡Ý99.90%
  • Insoluble in Dilute HNO3 — ¡Ü0.020%
  • Antimony and Tin (as Sn) — ¡Ü0.01%
  • Arsenic — ¡Ü5ppm
  • Iron — ¡Ü0.005%
  • Lead — ¡Ü0.005%
  • Manganese — ¡Ü0.001%
  • Phosphorus — ¡Ü0.001%
  • Silver — ¡Ü20ppm

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat