Fisher Chemical Copper, Laboratory Grade, 500 g

Representative image only.
Fisher Chemical Mfr # C431500 – Item # EW-88069-53
Consistent, high quality organics and fine chemicals at great savings
  • Reagents and solutions to meet your lab and research needs

$577.00USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeLaboratory
  • Empirical FormulaCu
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)500
  • Formula Weight63.55
  • PackagingGlass Bottle
  • DOT InformationCopper; Class 9, Packing Group III
  • Storage SymbolRed
  • UN NumberUN3077
  • CAS Number7440-50-8
  • Boiling Point C (° C)2595
  • Melting Point C (° C)1083
  • DescriptionCopper, Laboratory Grade, 500 g
Storage Code Red

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat