Fisher Chemical C536250 o-Cresol (Laboratory) (250g) CAS 95-48-7

Representative image only.
Acros Organics Mfr # C536250 – Item # EW-88006-32
  • Freezing Point — 29°-31°C
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeLaboratory
  • Empirical FormulaC7H8O
  • Linear FormulaCH3C6H4OH
  • ClassSolids
  • ChemistrySolutions
  • Synonyms2-Methyl Phenol; o-Cresylic Acid; o-Hydroxytoluene
  • FormSolid
  • Weight (g)250
  • Formula Weight108.14
  • PackagingGlass bottle
  • DOT InformationCresols, solid; Class 6.1 (8), Packing Group II
  • Storage SymbolWhite
  • Hazard SymbolCorrosive, Toxic
  • CAS Number95487
  • UN NumberUN3455
  • Descriptiono-Cresol (Laboratory) (250g) CAS 95-48-7
CorrosiveStorage Code WhiteToxic

Key Features

  • Freezing Point — 29°-31°C

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat