Fisher Chemical C556500 Cyclohexane (Certified ACS) (500mL)

Representative image only.
Fisher Chemical Mfr # C556500 – Item # EW-88006-54
Quality and consistency your lab can count on
  • Assay — ≥99.0%
  • Color (APHA) — ≤10
  • Density at 25°C — 0.772-0.776g/mL
  • Residue after Evaporation — ≤0.002%
  • Substances Darkened by H2SO4 — To pass test
  • Water — ≤0.02%
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • ChemistrySolvents;Analytical Reagents
  • Volume (mL)500
  • Formula Weight84.16
  • CAS Number110-82-7
  • DescriptionCyclohexane (Certified ACS) (500mL)
FlammableHarmfulStorage Code Red

Key Features

  • Assay — ≥99.0%
  • Color (APHA) — ≤10
  • Density at 25°C — 0.772-0.776g/mL
  • Residue after Evaporation — ≤0.002%
  • Substances Darkened by H2SO4 — To pass test
  • Water — ≤0.02%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat