Fisher Chemical Chloroform, in 0.75% Ethanol as Preservative, HPLC Grade, 4 L

Representative image only.
Fisher Chemical Mfr # C6064 – Item # EW-88005-28
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay (including preservative) — ≥99.8%
  • Color (APHA) — ≤10
  • Preservative (Ethanol) — 0.5-1.3 v/v%
  • Optical Absorbance at 244nm — ≤1.00
  • at 245nm — ≤0.80
  • at 250nm — ≤0.28
  • at 254nm — ≤0.13
  • Fluorescence Background (as Quinine Sulfate) — To pass test
  • Refractive Index at 25°C — 1.4420-1.4450
  • Acid and Chloride — To pass test
  • Free Chlorine — To pass test
  • Residue after Evaporation — ≤2ppm
  • Substances Darkened by H2SO4 — To pass test
  • Water — ≤0.02%
  • Acetone and Aldehyde [as (CH3)2CO] — To pass test (about 0.005%)
  • Density at 25°C — 1.471-1.476g/mL
  • Lead — ≤0.05ppm
  • Suitability for Use in Dithizone Tests — To pass test
  • Submicron Filtered.

PRODUCT OPTIONS

$450.00USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeHPLC
  • Concentration0.9925
  • MatrixEthanol
  • Empirical FormulaCHCl3
  • ClassReagent Solvents
  • ChemistrySolutions
  • FormLiquid
  • SynonymsFormyl trichloride, Methane trichloride, Methenyl trichloride
  • Volume (mL)4000
  • Formula Weight119.38
  • PackagingGlass Bottle
  • DOT InformationChloroform; Class 6.1, Packing Group III
  • Storage SymbolBlue
  • Hazard SymbolHarmful
  • CAS Number67-66-3
  • UN NumberUN1888
  • Boiling Point C (° C)61
  • Melting Point C (° C)-63
  • DescriptionChloroform, in 0.75% Ethanol as Preservative, HPLC Grade, 4 L
Harmful

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (including preservative) — ≥99.8%
  • Color (APHA) — ≤10
  • Preservative (Ethanol) — 0.5-1.3 v/v%
  • Optical Absorbance at 244nm — ≤1.00
  • at 245nm — ≤0.80
  • at 250nm — ≤0.28
  • at 254nm — ≤0.13
  • Fluorescence Background (as Quinine Sulfate) — To pass test
  • Refractive Index at 25°C — 1.4420-1.4450
  • Acid and Chloride — To pass test
  • Free Chlorine — To pass test
  • Residue after Evaporation — ≤2ppm
  • Substances Darkened by H2SO4 — To pass test
  • Water — ≤0.02%
  • Acetone and Aldehyde [as (CH3)2CO] — To pass test (about 0.005%)
  • Density at 25°C — 1.471-1.476g/mL
  • Lead — ≤0.05ppm
  • Suitability for Use in Dithizone Tests — To pass test
  • Submicron Filtered.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat