Fisher Chemical C6074 Chloroform, Approx. 50ppm Pentene as Preservative/HPLC, Glass Bottle; 4 L

Representative image only.
Fisher Chemical Mfr # C6074 – Item # EW-88005-34
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay (including preservative) — ≥99.8%
  • Color (APHA) — ≤10
  • Preservative (Pentene) — 50ppm
  • Optical Absorbance at 245nm — ≤1.00
  • at 250nm — ≤0.32
  • at 254nm — ≤0.15
  • at 280nm — ≤0.01
  • Fluorescence Background (as Quinine Sulfate) — To pass test
  • Refractive Index at 25°C — 1.4430-1.4460
  • Residue after Evaporation — ≤2ppm
  • Water — ≤0.02%
  • Acid and Chloride — To pass test
  • Free Chlorine — To pass test
  • Lead — ≤0.05ppm
  • Suitability for Use in Dithizone Tests — To pass test
  • Acetone and Aldehyde [as (CH3)2CO] — To pass test (about 0.005%)
  • Submicron Filtered.
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeHPLC
  • Empirical FormulaCHCl3
  • Linear FormulaCHCl3
  • ClassReagent Solvents
  • ChemistrySolutions
  • FormLiquid
  • SynonymsTrichloromethane
  • Volume (mL)4000
  • Formula Weight119.38
  • PackagingAmber Glass
  • DOT InformationChloroform; Class 6.1, Packing Group III
  • Storage SymbolBlue
  • Hazard SymbolHarmful
  • CAS Number67-66-3
  • UN NumberUN1888
  • DescriptionChloroform (Approx. 50ppm Pentene as Preservative/HPLC) (4 L)
Harmful

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (including preservative) — ≥99.8%
  • Color (APHA) — ≤10
  • Preservative (Pentene) — 50ppm
  • Optical Absorbance at 245nm — ≤1.00
  • at 250nm — ≤0.32
  • at 254nm — ≤0.15
  • at 280nm — ≤0.01
  • Fluorescence Background (as Quinine Sulfate) — To pass test
  • Refractive Index at 25°C — 1.4430-1.4460
  • Residue after Evaporation — ≤2ppm
  • Water — ≤0.02%
  • Acid and Chloride — To pass test
  • Free Chlorine — To pass test
  • Lead — ≤0.05ppm
  • Suitability for Use in Dithizone Tests — To pass test
  • Acetone and Aldehyde [as (CH3)2CO] — To pass test (about 0.005%)
  • Submicron Filtered.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat