Fisher Chemical C77500 Calcium Chloride (Desiccant, 20 Mesh and Finer/Certified) (500g)

Representative image only.
Fisher Chemical Mfr # C77500 – Item # EW-88004-36
  • Assay — ≥95.0%
  • Titratable Base — ≤0.006 mEq/g
  • Magnesium and Alkali Salts (as SO4) — ≤2.0%
  • Mesh Size:
  • Retention on U.S. No. 20 Sieve — ≤10%

PRODUCT OPTIONS

$386.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ClassReagent Salts
  • ChemistrySolutions
  • Weight (g)500
  • Formula Weight110.99
  • CAS Number10043-52-4
  • DescriptionCalcium Chloride (Desiccant, 20 Mesh and Finer/Certified) (500 g)

Key Features

  • Assay — ≥95.0%
  • Titratable Base — ≤0.006 mEq/g
  • Magnesium and Alkali Salts (as SO4) — ≤2.0%
  • Mesh Size:
  • Retention on U.S. No. 20 Sieve — ≤10%
  • Retention on U.S. No. 200 Sieve — ≤30%

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat