Fisher Chemical Copper (II) Sulfate Pentahydrate, Technical Grade, 500 g

Representative image only.
Fisher Chemical Mfr # C489500 – Item # EW-88006-26
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeTechnical
  • Empirical FormulaCuSO4•5H2O
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • Synonyms Blue vitriol
  • FormSolid
  • Weight (g)500
  • Formula Weight249.68
  • PackagingPoly Bottle
  • DOT InformationCopper (II) sulfate pentahydrate (1:1:5); Class 9, Packing Group III
  • Storage SymbolBlue
  • Hazard SymbolEnvironment, Harmful
  • CAS Number7758-99-8
  • UN NumberUN3077
  • Boiling Point C (° C)150
  • Melting Point C (° C)110
  • DescriptionCopper (II) Sulfate Pentahydrate, Technical Grade, 500 g
Harmful

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat