Fisher Chemical Dextrose Anhydrous, Certified ACS Grade, 500 g

Representative image only.
Fisher Chemical Mfr # D16500 – Item # EW-88007-14
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Specific Rotation [α]&alpha25D; — +52.5° to +53.0°
  • Insoluble Matter — ≤0.005%
  • Loss on Drying at105°C — ≤0.2%
  • Residue after Ignition — ≤0.02%
  • Titratable Acid — ≤0.002mEq/g
  • Chloride — ≤0.01%
  • Sulfate and Sulfite (as SO4) — ≤0.005%
  • Starch — To pass test
  • Heavy Metals (as Pb) — ≤5ppm
  • Iron — ≤5ppm

$168.75USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaCH2OH(CHOH)4CHO
  • ClassSolids
  • ChemistryAnalytical Reagents
  • SynonymsSugar Starch, Corn Sugar, Cerelose, Glucose
  • FormSolid
  • Weight (g)500
  • Formula Weight180.16
  • PackagingGlass Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolFlammable
  • CAS Number50-99-7
  • Melting Point C (° C)146.1
  • DescriptionDextrose Anhydrous, Certified ACS Grade, 500 g
Storage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Specific Rotation [α]&alpha25D; — +52.5° to +53.0°
  • Insoluble Matter — ≤0.005%
  • Loss on Drying at105°C — ≤0.2%
  • Residue after Ignition — ≤0.02%
  • Titratable Acid — ≤0.002mEq/g
  • Chloride — ≤0.01%
  • Sulfate and Sulfite (as SO4) — ≤0.005%
  • Starch — To pass test
  • Heavy Metals (as Pb) — ≤5ppm
  • Iron — ≤5ppm

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat