Fisher Chemical E17720 Ethylene Glycol (Laboratory) (20 liter)

Representative image only.
Cole-Parmer Mfr # E17720 – Item # EW-88072-81
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC2H6O2
  • Linear FormulaHOCH2CH2OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • Synonyms1,2-Ethanediol
  • Volume (mL)20000
  • Formula Weight62.07
  • CAS Number107-21-1
  • DescriptionEthylene Glycol (Laboratory) (20 L)
HarmfulStorage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat