Fisher Chemical Ethylene Glycol, Certified Grade, 4 L

Representative image only.
Fisher Chemical Mfr # E1784 – Item # EW-88008-94
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Color (APHA) — ≤15
  • Acidity (as Acetic Acid)—≤0.010%
  • Water—≤0.2%
  • Boiling Range—196°-199°C
  • Specific Gravity at 25°C—1.112-1.115

$645.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeCertified
  • Empirical FormulaHOCH2CH2OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • Synonyms1,2-Ethanediol
  • Volume (mL)4000
  • Formula Weight62.07
  • PackagingGlass Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolHarmful
  • CAS Number107-21-1
  • Boiling Point C (° C)196
  • Melting Point C (° C)-13
  • DescriptionEthylene Glycol, Certified Grade, 4 L
HarmfulStorage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Color (APHA) — ≤15
  • Acidity (as Acetic Acid)—≤0.010%
  • Water—≤0.2%
  • Boiling Range—196°-199°C
  • Specific Gravity at 25°C—1.112-1.115

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat