Fisher Chemical E178500 Ethylene Glycol (Certified) (500 mL) CAS 107-21-1

Representative image only.
Fisher Chemical Mfr # E178500 – Item # EW-88008-90
  • Color (APHA) — ≤15
  • Acidity (as Acetic Acid)—≤0.010%
  • Water—≤0.2%
  • Boiling Range—196°-199°C
  • Specific Gravity at 25°C—1.112-1.115

PRODUCT OPTIONS

$181.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeCertified
  • Empirical FormulaC2H6O2
  • Linear FormulaHOCH2CH2OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • Synonyms1,2-Ethanediol
  • Volume (mL)500
  • Formula Weight62.07
  • PackagingAmber Glass
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolHarmful
  • CAS Number107-21-1
  • DescriptionEthylene Glycol (Certified) (500 mL)

Key Features

  • Color (APHA) — ≤15
  • Acidity (as Acetic Acid)—≤0.010%
  • Water—≤0.2%
  • Boiling Range—196°-199°C
  • Specific Gravity at 25°C—1.112-1.115

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat