Fisher Chemical Glycine, BP/EP/JP/USP Grade, 12 kg

Representative image only.
Acros Organics Mfr # G4512 – Item # EW-88073-97
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeBP/EP/JP/USP
  • Empirical FormulaCH2(NH2)(COOH)
  • SynonymsGly, Aminoacetic acid, Aminoethanoic acid, Gyn-Hydralin
  • FormSolid
  • Weight (g)12000
  • Formula Weight75.07
  • PackagingPoly Pail
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number56-40-6
  • Melting Point C (° C)182
  • DescriptionGlycine, BP/EP/JP/USP Grade, 12 kg

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat