Fisher Chemical Ferrous Sulfate Heptahydrate, Certified ACS Grade, 10 kg

Representative image only.
Fisher Chemical Mfr # I14610 – Item # EW-88075-16
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeCertified ACS
  • Empirical FormulaFeSO4•7H2O
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • SynonymsIron (II) sulfate heptahydrate
  • Weight (g)10000
  • Formula Weight278.01
  • PackagingPoly Pail
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolFlammable, Harmful
  • CAS Number7782-63-0
  • Boiling Point C (° C)300
  • Melting Point C (° C)64
  • DescriptionFerrous Sulfate Heptahydrate, Certified ACS Grade, 10 kg
HarmfulStorage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat