Fisher Chemical Methyl Ethyl Ketone, ACS Reagent Grade, 500 mL

Representative image only.
Fisher Chemical Mfr # M209500 – Item # EW-88016-50
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay (by GC) — ≥99.0%
  • Color (APHA) — ≤15
  • Density at 25°C — 0.799-0.801g/mL
  • Residue after Evaporation — ≤0.0025%
  • Titratable Acid — ≤0.0005mEq/g
  • Water — ≤0.1%

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaC4H8O
  • ClassReagent Solvents
  • ChemistryAnalytical Reagents
  • FormLiquid
  • SynonymsEthyl Methyl Ketone, 2-Butanone
  • Volume (mL)500
  • Formula Weight72.11
  • PackagingGlass Bottle
  • DOT InformationEthyl methyl ketone; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable, Irritant
  • UN NumberUN1193
  • CAS Number78-93-3
  • Boiling Point C (° C)80
  • Melting Point C (° C)-87
  • DescriptionMethyl Ethyl Ketone, ACS Reagent Grade, 500 mL
FlammableIrritantStorage Code Red

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (by GC) — ≥99.0%
  • Color (APHA) — ≤15
  • Density at 25°C — 0.799-0.801g/mL
  • Residue after Evaporation — ≤0.0025%
  • Titratable Acid — ≤0.0005mEq/g
  • Water — ≤0.1%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat