Fisher Chemical Malachite Green Oxalate, Certified Biological Stain Grade, 25 g

Representative image only.
Fisher Chemical Mfr # M29025 – Item # EW-88076-30
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

$187.75USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeCertified Biological Stain
  • Empirical FormulaC23H25N2•C2HO4•0.5C2H2O4
  • ChemistrySolutions
  • FormSolid
  • SynonymsC.I. 42000, Basic Green 4
  • Weight (g)25
  • Formula Weight327.33
  • PackagingGlass Bottle
  • DOT InformationMalachite Green Oxalate; Class 6.1, Packing Group III
  • Storage SymbolBlue
  • CAS Number2437-29-8
  • UN NumberUN2811
  • Melting Point C (° C)164
  • DescriptionMalachite Green Oxalate, Certified Biological Stain Grade, 25 g
Harmful

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat